2-(Ethoxycarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-(Ethoxycarbonyl)benzeneboronic acid, (2-(Ethoxycarbonyl)phenyl)boronic acid

BB-2126-5G BB-2126-25G BB-2126-1G
COBBBB-2126-5GEA 63.4 EUR
COBBBB-2126-5G COBBBB-2126-25G COBBBB-2126-1G
2-(Ethoxycarbonyl)phenylboronsäure
2-(Ethoxycarbonyl)phenylboronsäure
Formel: C₉H₁₁BO₄
Molecular Weight: 193.99 g/mol
MDL: MFCD02179453
CAS-Nummer: 380430-53-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR