3-(Ethoxycarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-(Ethoxycarbonyl)benzeneboronic acid, 3-Ethoxycarbonylphenylboronic acid

BB-2117-25G BB-2117-1G BB-2117-5G BB-2117-100G
COBBBB-2117-25GEA 142 EUR
COBBBB-2117-25G COBBBB-2117-1G COBBBB-2117-5G COBBBB-2117-100G
3-(Ethoxycarbonyl)phenylboronsäure
3-(Ethoxycarbonyl)phenylboronsäure
Formel: C₉H₁₁BO₄
Molecular Weight: 193,99 g/mol
MDL: MFCD02179444
CAS-Nummer: 4334-87-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR