3-(Ethoxycarbonyl)phenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(Ethoxycarbonyl)benzeneboronic acid, 3-Ethoxycarbonylphenylboronic acid

M-Clarity™ Qualitätsstufe = MQ100

574651-1G 574651-5G
SIAL574651-1GEA 28.1 EUR
SIAL574651-1G SIAL574651-5G
3-(Ethoxycarbonyl)phenylboronsäure, Sigma-Aldrich®
3-(Ethoxycarbonyl)phenylboronsäure
Formel: C₉H₁₁BO₄
Molecular Weight: 193,99 g/mol
MDL: MFCD02179444
CAS-Nummer: 4334-87-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR