4-(Ethoxycarbonyl)phenylboronsäure ≥94%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-(Ethoxycarbonyl)benzeneboronic acid, (4-Ethoxycarbonylphenyl)boronic acid
H26356.03
H26356.06
H26356.03EA
29.9
EUR
H26356.03
H26356.06
4-(Ethoxycarbonyl)phenylboronsäure ≥94%
4-(Ethoxycarbonyl)phenylboronsäure
Purity: 94%
CAS No.: 4334-88-7
Molecular Formula: C9H11BO4
Formula Weight: 193.99
Melting Point: 137-140°
MDL No.: MFCD02179441
Formel:
C₉H₁₁BO₄ Molecular Weight: 193,99 g/mol Schmelzpunkt: 137…140 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02179441 CAS-Nummer: 4334-88-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...