4-(Ethoxycarbonyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
4-(Ethoxycarbonyl)benzeneboronic acid, (4-Ethoxycarbonylphenyl)boronic acid
BB-2113-5G
BB-2113-25G
BB-2113-1G
COBBBB-2113-5GEA
30.4
EUR
COBBBB-2113-5G
COBBBB-2113-25G
COBBBB-2113-1G
4-(Ethoxycarbonyl)phenylboronsäure
4-(Ethoxycarbonyl)phenylboronsäure
Formel:
C₉H₁₁BO₄ Molecular Weight: 193,99 g/mol |
MDL:
MFCD02179441 CAS-Nummer: 4334-88-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...