4-(Ethoxycarbonyl)phenylboronsäure

Lieferant: FLUOROCHEM

Synonyme: 4-(Ethoxycarbonyl)benzeneboronic acid, (4-Ethoxycarbonylphenyl)boronic acid

010792-5G 010792-25G
FLUO010792-5GEA 57.9 EUR
FLUO010792-5G FLUO010792-25G
4-(Ethoxycarbonyl)phenylboronsäure
4-(Ethoxycarbonyl)phenylboronsäure
Formel: C₉H₁₁BO₄
Molecular Weight: 193,99 g/mol
MDL: MFCD02179441
CAS-Nummer: 4334-88-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR