4-(Ethoxycarbonyl)-2-nitrophenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Ethoxycarbonyl-2-nitrophenylboronic acid, 4-Ethoxycarbonyl-2-nitrobenzeneboronic acid

BB-2148-5G BB-2148-1G
COBBBB-2148-5GEA 115 EUR
COBBBB-2148-5G COBBBB-2148-1G
4-(Ethoxycarbonyl)-2-nitrophenylboronsäure
4-(Ethoxycarbonyl)-2-nitrophenylboronsäure
Formel: C₉H₁₀BNO₆
Molecular Weight: 238.9898 g/mol
MDL: MFCD02179460
CAS-Nummer: 5785-70-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR