3-(Ethoxycarbonyl)-5-nitrophenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-Ethoxycarbonyl-5-nitrophenylboronic acid, (3-Ethoxycarbonyl-5-nitrophenyl)boronic acid
BB-2010-25G
BB-2010-5G
BB-2010-1G
COBBBB-2010-25GEA
471
EUR
COBBBB-2010-25G
COBBBB-2010-5G
COBBBB-2010-1G
3-(Ethoxycarbonyl)-5-nitrophenylboronsäure
3-(Ethoxycarbonyl)-5-nitrophenylboronsäure
Formel:
C₉H₁₀BNO₆ Molecular Weight: 238,9898 g/mol |
MDL:
MFCD03788422 CAS-Nummer: 850568-37-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...