2-Carboxyphenylboronsäure 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Boronobenzoic acid, 2-Carboxybenzeneboronic acid

L16301.03 L16301.06 L16301.MD
L16301.03EA 213 EUR
L16301.03 L16301.06 L16301.MD
2-Carboxyphenylboronsäure 95%
2-Carboxyphenylboronsäure

Purity: 95% CAS No.: 149105-19-1 Molecular Formula: C7H7BO4 Formula Weight: 165.94 Melting Point: ca 220° Beilstein Reference: 2835407

Formel: C₇H₇BO₄
Molecular Weight: 165,94 g/mol
Schmelzpunkt: ∼ 220 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01318118
CAS-Nummer: 149105-19-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR