2-Carboxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Boronobenzoic acid, 2-Carboxybenzeneboronic acid

BB-2469-1G BB-2469-5G BB-2469-25G
COBBBB-2469-1GEA 38.7 EUR
COBBBB-2469-1G COBBBB-2469-5G COBBBB-2469-25G
2-Carboxyphenylboronsäure
2-Carboxyphenylboronsäure
Formel: C₇H₇BO₄
Molecular Weight: 165,94 g/mol
MDL: MFCD01318118
CAS-Nummer: 149105-19-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR