2-Carboxyphenylboronsäure
Lieferant: FLUOROCHEM
Synonyme:
2-Boronobenzoic acid, 2-Carboxybenzeneboronic acid
012727-1G
012727-5G
FLUO012727-1GEA
48.8
EUR
FLUO012727-1G
FLUO012727-5G
2-Carboxyphenylboronsäure
2-Carboxyphenylboronsäure
Formel:
C₇H₇BO₄ Molecular Weight: 165,94 g/mol |
MDL:
MFCD01318118 CAS-Nummer: 149105-19-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...