2-Carboxyphenylboronsäure Dihydrat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

704784-1G
SIAL704784-1GEA 131 EUR
SIAL704784-1G
2-Carboxyphenylboronsäure Dihydrat, Sigma-Aldrich®
2-Carboxyphenylboronsäure Dihydrat
Formel: C₇H₇BO₄·2H₂O
MDL: MFCD15141948
CAS-Nummer: 1257650-88-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR