3-Carboxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Boronobenzoic acid, 3-Carboxybenzeneboronic acid

BB-2470-5G BB-2470-25G BB-2470-100G
COBBBB-2470-5GEA 26.2 EUR
COBBBB-2470-5G COBBBB-2470-25G COBBBB-2470-100G
3-Carboxyphenylboronsäure
3-Carboxyphenylboronsäure
Formel: C₇H₇BO₄
Molecular Weight: 165.94 g/mol
MDL: MFCD00036833
CAS-Nummer: 25487-66-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR