3-Carboxyphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Boronobenzoic acid, 3-Carboxybenzeneboronic acid

M-Clarity™ Qualitätsstufe = MQ100

456764-10G 456764-1G
SIAL456764-10GEA 77.4 EUR
SIAL456764-10G SIAL456764-1G
3-Carboxyphenylboronsäure, Sigma-Aldrich®
3-Carboxyphenylboronsäure
Formel: C₇H₇BO₄
Molecular Weight: 165,94 g/mol
MDL: MFCD00036833
CAS-Nummer: 25487-66-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR