4-Carboxyphenylboronsäure ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Boronobenzoic Acid, Ark Pharm, 4-Carboxybenzeneboronic acid

B20954.03 B20954.06 B20954.14
B20954.03EA 34.7 EUR
B20954.03 B20954.06 B20954.14
4-Carboxyphenylboronsäure ≥97%
4-Carboxyphenylboronsäure

Purity: 97% CAS No.: 14047-29-1 Molecular Formula: C7H7BO4 Formula Weight: 165.94 Melting Point: ca 220° dec. Beilstein Reference: 3031088

Formel: C₇H₇BO₄
Molecular Weight: 165,94 g/mol
Schmelzpunkt: ∼ 220 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00151801
CAS-Nummer: 14047-29-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR