4-Carboxyphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
4-Boronobenzoic Acid, Ark Pharm, 4-Carboxybenzeneboronic acid
BB-2255-5G
BB-2255-25G
BB-2255-100G
COBBBB-2255-5GEA
25.5
EUR
COBBBB-2255-5G
COBBBB-2255-25G
COBBBB-2255-100G
4-Carboxyphenylboronsäure
4-Carboxyphenylboronsäure
Formel:
C₇H₇BO₄ Molecular Weight: 165,94 g/mol |
MDL:
MFCD00151801 CAS-Nummer: 14047-29-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...