4-Carboxyphenylboronsäure

Lieferant: CHEMPUR

Synonyme: 4-Boronobenzoic Acid, Ark Pharm, 4-Carboxybenzeneboronic acid

FL10786.2 010786-5G 010786-25G
CHMPFL10786.2EA 0 EUR
CHMPFL10786.2 FLUO010786-5G FLUO010786-25G
4-Carboxyphenylboronsäure
4-Carboxyphenylboronsäure
Formel: C₇H₇BO₄
Molecular Weight: 165,94 g/mol
MDL: MFCD00151801
CAS-Nummer: 14047-29-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR