4-Carboxyphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Boronobenzoic Acid, Ark Pharm, 4-Carboxybenzeneboronic acid

M-Clarity™ Qualitätsstufe = MQ200

456772-10G 456772-1G
SIAL456772-10GEA 343 EUR
SIAL456772-10G SIAL456772-1G
4-Carboxyphenylboronsäure, Sigma-Aldrich®
4-Carboxyphenylboronsäure
Formel: C₇H₇BO₄
Molecular Weight: 165,94 g/mol
MDL: MFCD00151801
CAS-Nummer: 14047-29-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR