4-Carboxy-2-nitrophenylboronsäure
Lieferant: COMBI-BLOCKS
BB-2451-5G
BB-2451-1G
COBBBB-2451-5GEA
0
EUR
COBBBB-2451-5G
COBBBB-2451-1G
4-Carboxy-2-nitrophenylboronsäure
4-Carboxy-2-nitrophenylboronsäure
Formel:
C₇H₆BNO₆ |
MDL:
MFCD00448780 CAS-Nummer: 85107-54-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...