3-Carboxy-5-nitrophenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Carboxy-5-nitrobenzeneboronic acid, 3-Borono-5-nitrobenzoic acid

BB-2476-25G BB-2476-5G BB-2476-1G
COBBBB-2476-25GEA 193 EUR
COBBBB-2476-25G COBBBB-2476-5G COBBBB-2476-1G
3-Carboxy-5-nitrophenylboronsäure
3-Carboxy-5-nitrophenylboronsäure
Formel: C₇H₆BNO₆
Molecular Weight: 210.94 g/mol
MDL: MFCD00757433
CAS-Nummer: 101084-81-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR