3-Carboxy-5-nitrophenylboronsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3-Carboxy-5-nitrobenzeneboronic acid, 3-Borono-5-nitrobenzoic acid
721042-1G
721042-5G
SIAL721042-1GEA
30.7
EUR
SIAL721042-1G
SIAL721042-5G
3-Carboxy-5-nitrophenylboronsäure, Sigma-Aldrich®
3-Carboxy-5-nitrophenylboronsäure
Formel:
C₇H₆BNO₆ Molecular Weight: 210,94 g/mol |
MDL:
MFCD00757433 CAS-Nummer: 101084-81-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...