3-(N,N-Dimethylamino)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-Dimethylaminophenylboronic acid, 3-(Dimethylamino)benzeneboronic acid
BB-8332-5G
BB-8332-1G
COBBBB-8332-5GEA
98.3
EUR
COBBBB-8332-5G
COBBBB-8332-1G
3-(N,N-Dimethylamino)phenylboronsäure
3-(N,N-Dimethylamino)phenylboronsäure
Formel:
C₈H₁₂BNO₂ Molecular Weight: 164.9974 g/mol |
MDL:
MFCD01318996 CAS-Nummer: 178752-79-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...