3-Aminophenylboronsäure Hemisulfat ≥98%

Lieferant: Thermo Scientific

Synonyme: (3-Aminophenyl)boronic acid sulphate (2:1), 3-Aminobenzeneboronic acid hemisulphate

AC10430.1 AC10430.10
MAYBAC10430.1EA 0 EUR
MAYBAC10430.1 MAYBAC10430.10
3-Aminophenylboronsäure Hemisulfat ≥98%
3-Aminophenylboronsäure Hemisulfat
Formel: C₁₂H₁₈B₂N₂O₈S
Molecular Weight: 371,97 g/mol
MDL: MFCD00013111
CAS-Nummer: 66472-86-4
EINECS: 266-376-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR