3-Aminophenylboronsäure Hemisulfat

Lieferant: COMBI-BLOCKS

Synonyme: (3-Aminophenyl)boronic acid sulphate (2:1), 3-Aminobenzeneboronic acid hemisulphate

BB-8300-1G BB-8300-5G
COBBBB-8300-1GEA 55.7 EUR
COBBBB-8300-1G COBBBB-8300-5G
3-Aminophenylboronsäure Hemisulfat
3-Aminophenylboronsäure Hemisulfat
Formel: C₁₂H₁₈B₂N₂O₈S
Molecular Weight: 371.97 g/mol
MDL: MFCD00013111
CAS-Nummer: 66472-86-4
EINECS: 266-376-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR