3-Aminophenylboronsäure Hemisulfat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (3-Aminophenyl)boronic acid sulphate (2:1), 3-Aminobenzeneboronic acid hemisulphate

M-Clarity™ Qualitätsstufe = MQ200

A71751-25G A71751-5G
SIALA71751-25GEA 372 EUR
SIALA71751-25G SIALA71751-5G
3-Aminophenylboronsäure Hemisulfat, Sigma-Aldrich®
3-Aminophenylboronsäure Hemisulfat
Formel: C₁₂H₁₈B₂N₂O₈S
Molecular Weight: 371.97 g/mol
MDL: MFCD00013111
CAS-Nummer: 66472-86-4
EINECS: 266-376-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR