2-Aminophenylboronsäure Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

721379-5G 721379-1G
SIAL721379-5GEA 234 EUR
SIAL721379-5G SIAL721379-1G
2-Aminophenylboronsäure Hydrochlorid, Sigma-Aldrich®
2-Aminophenylboronsäure Hydrochlorid
Formel: C₆H₉BClNO₂
Molecular Weight: 173.4052 g/mol
MDL: MFCD02258096
CAS-Nummer: 863753-30-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR