3-Aminophenylboronsäure Hydrochlorid

Lieferant: COMBI-BLOCKS

Synonyme: (3-Aminophenyl)boronic acid hydrochloride, 3-Aminobenzeneboronic acid hydrochloride

BB-2229-5G BB-2229-25G BB-2229-1G
COBBBB-2229-5GEA 152 EUR
COBBBB-2229-5G COBBBB-2229-25G COBBBB-2229-1G
3-Aminophenylboronsäure Hydrochlorid
3-Aminophenylboronsäure Hydrochlorid
Formel: C₆H₉BClNO₂
Molecular Weight: 173,41 g/mol
MDL: MFCD00191748
CAS-Nummer: 85006-23-1
EINECS: 285-052-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR