3-Aminophenylboronsäure Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (3-Aminophenyl)boronic acid hydrochloride, 3-Aminobenzeneboronic acid hydrochloride

M-Clarity™ Qualitätsstufe = MQ100

410705-1G 410705-5G
SIAL410705-1GEA 88.4 EUR
SIAL410705-1G SIAL410705-5G
3-Aminophenylboronsäure Hydrochlorid, Sigma-Aldrich®
3-Aminophenylboronsäure Hydrochlorid
Formel: C₆H₉BClNO₂
Molecular Weight: 173,41 g/mol
MDL: MFCD00191748
CAS-Nummer: 85006-23-1
EINECS: 285-052-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR