4-Aminophenylboronsäure Hydrochlorid

Lieferant: COMBI-BLOCKS

Synonyme: 4-Aminobenzeneboronic acid hydrochloride, (4-Aminophenyl)boronic acid hydrochloride

BB-2230-5G BB-2230-1G
COBBBB-2230-5GEA 76.7 EUR
COBBBB-2230-5G COBBBB-2230-1G
4-Aminophenylboronsäure Hydrochlorid
4-Aminophenylboronsäure Hydrochlorid
Formel: C₆H₉BClNO₂
Molecular Weight: 173.41 g/mol
MDL: MFCD03001333
CAS-Nummer: 80460-73-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR