4-Brom-5-fluor-2-nitroanilin

Lieferant: COMBI-BLOCKS

AN-1501-5G AN-1501-1G
COBBAN-1501-5GEA 462 EUR
COBBAN-1501-5G COBBAN-1501-1G
4-Brom-5-fluor-2-nitroanilin
4-Brom-5-fluor-2-nitroanilin
Formel: C₆H₄BrFN₂O₂
Molecular Weight: 235,0106 g/mol
MDL: MFCD11101433
CAS-Nummer: 153505-36-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR