4-Carboxy-3-chlorphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-Carboxy-3-chlorophenylboronic acid, pinacol ester, 2-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

PN-2745-5G PN-2745-25G PN-2745-1G
COBBPN-2745-5GEA 548 EUR
COBBPN-2745-5G COBBPN-2745-25G COBBPN-2745-1G
4-Carboxy-3-chlorphenylboronsäurepinakolester
4-Carboxy-3-chlorphenylboronsäurepinakolester
Formel: C₁₃H₁₆BClO₄
Molecular Weight: 282,5277 g/mol
MDL: MFCD12546621
CAS-Nummer: 890839-31-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR