2-Chlor-(4-methoxycarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chloro-4-(methoxycarbonyl)phenylboronic acid, 2-Chloro-4-(methoxycarbonyl)benzeneboronic acid, (2-Chloro-4-(methoxycarbonyl)phenyl)boronic acid

BB-3530-5G BB-3530-1G
COBBBB-3530-5GEA 298 EUR
COBBBB-3530-5G COBBBB-3530-1G
2-Chlor-(4-methoxycarbonyl)phenylboronsäure
2-Chlor-(4-methoxycarbonyl)phenylboronsäure
Formel: C₈H₈BClO₄
Molecular Weight: 214,4107 g/mol
MDL: MFCD08689530
CAS-Nummer: 603122-80-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR