3-Chlor-(4-ethoxycarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-Chloro-4-(ethoxycarbonyl)phenyl)boronic acid, 3-Chloro-4-(ethoxycarbonyl)phenylboronic acid

BB-2753-5G BB-2753-1G
COBBBB-2753-5GEA 646 EUR
COBBBB-2753-5G COBBBB-2753-1G
3-Chlor-(4-ethoxycarbonyl)phenylboronsäure
3-Chlor-(4-ethoxycarbonyl)phenylboronsäure
Formel: C₉H₁₀BClO₄
Molecular Weight: 228.4373 g/mol
MDL: MFCD06659875
CAS-Nummer: 850568-11-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR