3-Chlor-(4-methoxycarbonyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(3-Chloro-4-(methoxycarbonyl)phenyl)boronic acid, 3-Chloro-4-(methoxycarbonyl)phenylboronic acid, 3-Chloro-4-(methoxycarbonyl)benzeneboronic acid
BB-2752-5G
BB-2752-1G
COBBBB-2752-5GEA
221
EUR
COBBBB-2752-5G
COBBBB-2752-1G
3-Chlor-(4-methoxycarbonyl)phenylboronsäure
3-Chlor-(4-methoxycarbonyl)phenylboronsäure
Formel:
C₈H₈BClO₄ Molecular Weight: 214,4100 g/mol |
MDL:
MFCD06659874 CAS-Nummer: 603122-82-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...