4-Chlor-(2-ethoxycarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Chloro-2-ethoxycarbonylphenylboronic acid

BB-2183-5G BB-2183-1G
COBBBB-2183-5GEA 1690 EUR
COBBBB-2183-5G COBBBB-2183-1G
4-Chlor-(2-ethoxycarbonyl)phenylboronsäure
4-Chlor-(2-ethoxycarbonyl)phenylboronsäure
Formel: C₉H₁₀BClO₄
Molecular Weight: 228,4400 g/mol
MDL: MFCD06659823
CAS-Nummer: 850568-61-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR