4-Chlor-(3-ethoxycarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Chloro-3-(ethoxycarbony)phenylboronic acid, (4-Chloro-3-(ethoxycarbonyl)phenyl)boronic acid

BB-2852-1G BB-2852-5G
COBBBB-2852-1GEA 249 EUR
COBBBB-2852-1G COBBBB-2852-5G
4-Chlor-(3-ethoxycarbonyl)phenylboronsäure
4-Chlor-(3-ethoxycarbonyl)phenylboronsäure
Formel: C₉H₁₀BClO₄
Molecular Weight: 228.4400 g/mol
MDL: MFCD08235062
CAS-Nummer: 874219-46-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR