4-Chlor-(3-methoxycarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Chloro-3-(methoxycarbonyl)benzeneboronic acid, 4-Chloro-3-(methoxycarbonyl)phenylboronic acid

BB-2851-5G BB-2851-1G
COBBBB-2851-5GEA 0 EUR
COBBBB-2851-5G COBBBB-2851-1G
4-Chlor-(3-methoxycarbonyl)phenylboronsäure
4-Chlor-(3-methoxycarbonyl)phenylboronsäure
Formel: C₈H₈BClO₄
MDL: MFCD06801690
CAS-Nummer: 874219-45-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR