5-Chlor-(2-ethoxycarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-Chloro-2-(ethoxycarbonyl)benzeneboronic acid, (5-Chloro-2-(ethoxycarbonyl)phenyl)boronic acid

BB-2270-5G BB-2270-1G
COBBBB-2270-5GEA 1580 EUR
COBBBB-2270-5G COBBBB-2270-1G
5-Chlor-(2-ethoxycarbonyl)phenylboronsäure
5-Chlor-(2-ethoxycarbonyl)phenylboronsäure
Formel: C₉H₁₀BClO₄
Molecular Weight: 228.44 g/mol
MDL: MFCD07363744
CAS-Nummer: 871329-55-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR