5-Chlor-(2-methoxycarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (5-Chloro-2-(methoxycarbonyl)phenyl)boronic acid

BB-5074-1G BB-5074-5G
COBBBB-5074-1GEA 375 EUR
COBBBB-5074-1G COBBBB-5074-5G
5-Chlor-(2-methoxycarbonyl)phenylboronsäure
5-Chlor-(2-methoxycarbonyl)phenylboronsäure
Formel: C₈H₈BClO₄
Molecular Weight: 214,4100 g/mol
MDL: MFCD09878336
CAS-Nummer: 957062-64-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR