3-Chlor-(5-methoxycarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Chloro-5-(methoxycarbonyl)phenylboronic acid, (3-Chloro-5-(methoxycarbonyl)phenyl)boronic acid

BB-3254-5G BB-3254-1G
COBBBB-3254-5GEA 0 EUR
COBBBB-3254-5G COBBBB-3254-1G
3-Chlor-(5-methoxycarbonyl)phenylboronsäure
3-Chlor-(5-methoxycarbonyl)phenylboronsäure
Formel: C₈H₈BClO₄
Molecular Weight: 214.4107 g/mol
MDL: MFCD09475906
CAS-Nummer: 957120-26-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR