4-Carboxy-2-chlorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Carboxy-2-chlorobenzeneboronic acid

BB-2746-5G BB-2746-25G BB-2746-1G
COBBBB-2746-5GEA 141 EUR
COBBBB-2746-5G COBBBB-2746-25G COBBBB-2746-1G
4-Carboxy-2-chlorphenylboronsäure
4-Carboxy-2-chlorphenylboronsäure
Formel: C₇H₆BClO₄
Molecular Weight: 200.39 g/mol
MDL: MFCD07363763
CAS-Nummer: 851335-09-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR