4-Carboxy-3-chlorphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
4-Carboxy-3-chlorobenzeneboronic acid
BB-2745-1G
BB-2745-5G
BB-2745-25G
COBBBB-2745-1GEA
37.4
EUR
COBBBB-2745-1G
COBBBB-2745-5G
COBBBB-2745-25G
4-Carboxy-3-chlorphenylboronsäure
4-Carboxy-3-chlorphenylboronsäure
Formel:
C₇H₆BClO₄ Molecular Weight: 200,39 g/mol |
MDL:
MFCD06656268 CAS-Nummer: 136496-72-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...