2-Carboxy-4-chlorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Carboxy-4-chlorobenzeneboronic acid

BB-2738-5G BB-2738-1G
COBBBB-2738-5GEA 1140 EUR
COBBBB-2738-5G COBBBB-2738-1G
2-Carboxy-4-chlorphenylboronsäure
2-Carboxy-4-chlorphenylboronsäure
Formel: C₇H₆BClO₄
Molecular Weight: 200.39 g/mol
MDL: MFCD06659869
CAS-Nummer: 850568-07-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR