2-Carboxy-5-chlorphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2-Borono-4-chlorobenzoic acid
BB-2747-5G
BB-2747-1G
COBBBB-2747-5GEA
1880
EUR
COBBBB-2747-5G
COBBBB-2747-1G
2-Carboxy-5-chlorphenylboronsäure
2-Carboxy-5-chlorphenylboronsäure
Formel:
C₇H₆BClO₄ Molecular Weight: 200.384 g/mol |
MDL:
MFCD08235091 CAS-Nummer: 874290-67-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...