3-Carboxy-5-chlorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Borono-5-chlorobenzoic acid

BB-3885-5G BB-3885-1G
COBBBB-3885-5GEA 737 EUR
COBBBB-3885-5G COBBBB-3885-1G
3-Carboxy-5-chlorphenylboronsäure
3-Carboxy-5-chlorphenylboronsäure
Formel: C₇H₆BClO₄
Molecular Weight: 200.3841 g/mol
MDL: MFCD09258748
CAS-Nummer: 957061-05-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR