5-Carboxy-2-chlorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chloro-5-carboxyphenylboronic acid

BB-3487-5G BB-3487-1G
COBBBB-3487-5GEA 165 EUR
COBBBB-3487-5G COBBBB-3487-1G
5-Carboxy-2-chlorphenylboronsäure
5-Carboxy-2-chlorphenylboronsäure
Formel: C₇H₆BClO₄
Molecular Weight: 200.3800 g/mol
MDL: MFCD08689491
CAS-Nummer: 913835-75-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR