4-Carboxy-2-fluorphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-Carboxy-2-fluorophenylboronic Acid Pinacol Ester, 4-Carboxy-2-fluorobenzeneboronic acid pinacol ester

PN-3230-1G PN-3230-5G
COBBPN-3230-1GEA 96.1 EUR
COBBPN-3230-1G COBBPN-3230-5G
4-Carboxy-2-fluorphenylboronsäurepinakolester
4-Carboxy-2-fluorphenylboronsäurepinakolester
Formel: C₁₃H₁₆BFO₄
Molecular Weight: 266,07 g/mol
MDL: MFCD12756466
CAS-Nummer: 1050423-87-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR