4-Carboxy-3-fluorphenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
PN-2632-1G
PN-2632-5G
COBBPN-2632-1GEA
149
EUR
COBBPN-2632-1G
COBBPN-2632-5G
4-Carboxy-3-fluorphenylboronsäurepinakolester
4-Carboxy-3-fluorphenylboronsäurepinakolester
Formel:
C₁₃H₁₆BFO₄ Molecular Weight: 266,0732 g/mol |
MDL:
MFCD09027284 CAS-Nummer: 867256-77-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...