3-Carboxy-4-fluorphenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid, 3-Carboxy-4-fluorobenzeneboronic acid pinacol ester
PN-2879-5G
PN-2879-1G
COBBPN-2879-5GEA
125
EUR
COBBPN-2879-5G
COBBPN-2879-1G
3-Carboxy-4-fluorphenylboronsäurepinakolester
3-Carboxy-4-fluorphenylboronsäurepinakolester
Formel:
C₁₃H₁₆BFO₄ Molecular Weight: 266,07 g/mol |
MDL:
MFCD12546519 CAS-Nummer: 882679-10-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...