3-Carboxy-5-fluorphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 3-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid, 3-Carboxy-5-fluorobenzeneboronic acid pinacol ester

PN-2797-5G PN-2797-1G
COBBPN-2797-5GEA 460 EUR
COBBPN-2797-5G COBBPN-2797-1G
3-Carboxy-5-fluorphenylboronsäurepinakolester
3-Carboxy-5-fluorphenylboronsäurepinakolester
Formel: C₁₃H₁₆BFO₄
Molecular Weight: 266,07 g/mol
MDL: MFCD12756465
CAS-Nummer: 936728-22-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR